missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Büchner Funnel with Fritted Disc

Top edge is beaded.

$69.20 - $76.65

Specifications

Certifications/Compliance ASTM E128
Length (English) Stem 2.95 in.
Length (Metric) Stem 75mm
Material Pyrex™
Pore Size 40 to 60μm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Diameter (English) Disc Diameter (English) Outer Stem Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Disc Diameter (English) Outer Stem Height (English) Price Quantity  
S34591-3 Fisher Science Education
3606030CEMD
1.01 oz. 1.18 in. 0.31 in. 5.47 in. Each for $69.20
Case of 6 Each for $415.15
S34591-4 Fisher Science Education
3606060CEMD
2.02 oz. 1.57 in. 0.39 in. 6.29 in. Each for $76.65
Case of 6 Each for $459.70
Specifications

Description

These funnels are particularly useful where the filter paper would be attacked by the material being filtered.

Description & Specifications

Specifications

ASTM E128
75mm
40 to 60μm
Useful where filter paper would be attacked by material being filtered
Coarse
2.95 in.
Pyrex™
Buchner Funnel
Fritted discs