PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts
For use with Accelerated One-Step Extractor/Concentrator Systems
Manufacturer:  PYREX™ C391847BP
 View more versions of this product
Catalog No. 06-423-1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Replacement part for Pyrex Accelerated One-Step extractor/concentrator system. The system performs extraction, concentration, and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
Specifications
Specifications
4.7 cm
Pyrex™ accelerated One-Step™ extractor or concentrator system
PTFE
47 mm PTFE Membrane, Bulk Pack
1.85 in.