Corning™ PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts
For use with Accelerated One-Step Extractor/Concentrator Systems
Supplier: Corning™ 3915MBO
Description
Replacement part for Pyrex Accelerated One-Step extractor/concentrator system. The system performs extraction, concentration, and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.Specifications
Extractor Body | |
Pyrex Accelerated One-Step Extractor or Concentrator System | |
45/50 |