PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts
For use with Accelerated One-Step Extractor/Concentrator Systems
Manufacturer: PYREX™ C3915CW
Description
Replacement part for Pyrex Accelerated One-Step extractor/concentrator system. The system performs extraction, concentration, and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.Specifications
Washer and Connector | |
Pyrex™ accelerated One-Step™ extractor or concentrator system |