missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ 47mm Microfiltration Glassware Replacement Parts

Replacement Parts for Corning PYREX 47mm Microfiltration Glassware Apparatus (13-645-088, sold separately)

Manufacturer:  Corning Incorporated C33971300

 View more versions of this product

Catalog No. 13-645-089



Specifications

Specifications

Graduated Funnel
75mm
PYREX 47mm Microfiltration Glassware Apparatus
Pyrex Glass
115mm
300mL
Documents