missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ 47mm Microfiltration Glassware Replacement Parts

Replacement Parts for Corning PYREX 47mm Microfiltration Glassware Apparatus (13-645-088, sold separately)

Manufacturer:  Corning Incorporated C339854L

 View more versions of this product

Catalog No. 13-645-087



Specifications

Specifications

Erlenmeyer Flask with 40/35 Standard Taper Joint, without tubulation
58mm
PYREX 47mm Microfiltration Glassware Apparatus
40/35
Pyrex Glass
120mm
4000mL
Documents