missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ 47mm Microfiltration Glassware Replacement Parts

Replacement Parts for Corning PYREX 47mm Microfiltration Glassware Apparatus (13-645-088, sold separately)

Manufacturer:  Corning Incorporated C33982B

 View more versions of this product

Catalog No. 13-645-085



Specifications

Specifications

Fritted support base w/tubulation
58mm
PYREX 47mm Microfiltration Glassware Apparatus
Glass
120mm
Documents