missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ 24/29 Standard Taper Stopcock Vacuum-Rated Glass Desiccators, Complete (Bowl and Cover)

Desiccators have a rugged 24/29 standard taper stopcock with a tooled hose connection that serves as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum.

Manufacturer:  Corning Incorporated C3121200

 View more versions of this product

Catalog No. 08-624-422


 

Alert:

Do not heat or subject to pressure. Rated to 1-atmosphere vacuum.


Specifications Description & Specifications

Specifications

1.53 gal.
5.8L
1cm
1.53 gal., 5.8L
Cover, bowl and stopcock, 24/29 standard taper external sleeves with tooled hose connections
Each
Large
Desiccator
29.6cm
11.5cm
Borosilicate Glass
22.3cm
19cm
4.5 in.
0.39 in.
PTFE
11.6 in.
Accessories