missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Class A Buret with Product Standard PTFE Stopcock Plug

Buret has colored markings, fine, sharp lines and large easy-to-read numbers

Manufacturer:  Corning Incorporated C2103100

 View more versions of this product

Catalog No. 03-701-25D


 

Specifications Description & Specifications

Specifications

3.38 oz.
A
1.8cm
75.2cm
Pyrex™
±0.1mL
3.38 oz., 100mL
0.2mL
ASTM E287, ASTM E542
0.70 in.
29.6 in.
Dust cap, colored markings, No. 2 PTFE fluorocarbon-resin stopcock
PTFE
100mL
Buret, 1/Cs.
Accessories