missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC432720010

 View more versions of this product

Catalog No. AC432720010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pyrazolo[1, 5-a]pyrimidine-5-carboxylic acid
Glass bottle
1g
97%
MFCD09414725
SBDWLDBGGVPVAR-UHFFFAOYSA-N
pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
28875802
Powder
Authentic
Yellow
1086375-50-9
C7H5N3O2
pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
C1=CN2C(=CC=N2)N=C1C(=O)O
163.136
163.14
≥96.0%

Chemical Identifiers

1086375-50-9
163.136
SBDWLDBGGVPVAR-UHFFFAOYSA-N
28875802
C1=CN2C(=CC=N2)N=C1C(=O)O
C7H5N3O2
MFCD09414725
pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS P Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents