Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 97%

$107.43 - $372.18

Chemical Identifiers

Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
CAS 25940-35-6
Molecular Formula C7H5N3O2
Formula Weight 163.14
MDL Number MFCD01975194
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5031803

  SDS
alfa aesar™
H5031803
1g Each for $372.18

AAH50318MD

  SDS
alfa aesar™
H50318MD
250mg Each for $107.43
Description & Specifications

Specifications

Formula Weight 163.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈180°C
Quantity 1g
CAS 25940-35-6
Health Hazard 1 Warning
Assay 97%
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
MDL Number MFCD01975194
Molecular Formula C7H5N3O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, is used as a chemicals, pharmaceutical intermediates, reagents. It is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.