Please login to your online account to display your discounted pricing

Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
CAS 378211-85-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5N3O2
Formula Weight 163.14
MDL Number MFCD00297221
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC434340050

  SDS
thermo fisher scientific
AC434340050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434340010

  SDS
thermo fisher scientific
AC434340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 163.14
Physical Form Powder or Crystalline Powder
Quantity 5g
Color White to orange
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
Molecular Formula C7H5N3O2
Assay Percent Range 97%
MDL Number MFCD00297221
CAS 378211-85-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.