Learn More
Pyrazinecarbonitrile, 99%, Thermo Scientific Chemicals
$211.91 - $211.91
Chemical Identifiers
CAS | 19847-12-2 |
---|---|
Molecular Formula | C5H3N3 |
Molecular Weight (g/mol) | 105.10 |
MDL Number | MFCD00049361 |
InChI Key | PMSVVUSIPKHUMT-UHFFFAOYSA-N |
Synonym | pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine |
PubChem CID | 73172 |
ChEBI | CHEBI:3982 |
IUPAC Name | pyrazine-2-carbonitrile |
SMILES | N#CC1=CN=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC436920500
|
Thermo Scientific Chemicals
436920500 |
50 g | Glass bottle |
Each for $211.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19847-12-2 | |
105.10 | |
PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
73172 | |
pyrazine-2-carbonitrile |
C5H3N3 | |
MFCD00049361 | |
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
CHEBI:3982 | |
N#CC1=CN=CC=N1 |
Specifications
19847-12-2 | |
Colorless to Yellow | |
90°C to 93°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5330 to 1.5350 (20°C, 589nm) | |
50 g | |
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
pyrazine-2-carbonitrile | |
73172 | |
105.1 | |
Liquid |
20°C | |
1.1740g/mL | |
97°C | |
98.5% min. (GC) | |
C5H3N3 | |
MFCD00049361 | |
1.174 | |
Solubility in water: soluble | |
N#CC1=CN=CC=N1 | |
105.10 | |
CHEBI:3982 | |
99% | |
Pyrazinecarbonitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 243-369-5
RUO – Research Use Only