Learn More
Pyrazine, 99+% (dry wt.) water <1.0%, Thermo Scientific Chemicals
$115.16 - $115.16
Chemical Identifiers
CAS | 290-37-9 |
---|---|
Molecular Formula | C4H4N2 |
Molecular Weight (g/mol) | 80.09 |
MDL Number | MFCD00006122 |
InChI Key | KYQCOXFCLRTKLS-UHFFFAOYSA-N |
Synonym | 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; |
PubChem CID | 9261 |
ChEBI | CHEBI:30953 |
IUPAC Name | pyrazine |
SMILES | C1=CN=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1398214
|
Thermo Scientific Chemicals
A1398214 |
25 g |
Each for $115.16
|
|
|||||
Description
Pyrazine is used in flavoring, fragrance, and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrazine is used in flavoring, fragrance, and dyes.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
290-37-9 | |
80.09 | |
KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
9261 | |
pyrazine |
C4H4N2 | |
MFCD00006122 | |
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
CHEBI:30953 | |
C1=CN=CC=N1 |
Specifications
290-37-9 | |
1.031 | |
55°C (131°F) | |
C4H4N2 | |
MFCD00006122 | |
UN1325 | |
Hygroscopic | |
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
pyrazine | |
9261 | |
80.09 | |
Pyrazine |
50°C to 56°C | |
113°C to 116°C | |
Strong | |
1.5235 | |
25 g | |
103905 | |
14,7957 | |
Soluble in water. | |
C1=CN=CC=N1 | |
80.09 | |
CHEBI:30953 | |
≥99% | |
(dry wt.) water <1.0% |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 206-027-6
RTECSNumber : UQ2015000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only