missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrazine, 98%

Manufacturer:  Alfa Aesar™ 1632609

 View more versions of this product

Catalog No. AA1632609



Specifications

Specifications

Pyrazine
55° (131°F)
Hygroscopic
White
10g
1.5235
C4H4N2
UN1325
14,7957
Soluble in water, alcohol, and ether
C1=CN=CC=N1
80.09
CHEBI:30953
Crystalline
1.031g/mL
Strong
113°C to 116°C
50°C to 56°C
290-37-9
98%
MFCD00006122
103905
1,4-Diazine
KYQCOXFCLRTKLS-UHFFFAOYSA-N
pyrazine
9261
80.09

Chemical Identifiers

290-37-9
80.09
KYQCOXFCLRTKLS-UHFFFAOYSA-N
9261
pyrazine
C4H4N2
MFCD00006122
1,4-Diazine
CHEBI:30953
C1=CN=CC=N1
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UQ2015000

TSCA : TSCA

SDS Documents