Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrazine-2-thiocarboxamide, 97%

$81.51 - $599.69

Chemical Identifiers

Chemical Name or Material Pyrazine-2-thiocarboxamide
CAS 4604-72-2
Molecular Formula C5H5N3S
Formula Weight 139.17
MDL Number MFCD00173663
Synonym Pyrazine-2-carbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2675003

  SDS
alfa aesar™
H2675003
1g Each for $198.89

AAH2675006

  SDS
alfa aesar™
H2675006
5g Each for $599.69

AAH26750MD

  SDS
alfa aesar™
H26750MD
250mg Each for $81.51
Description & Specifications

Specifications

Formula Weight 139.17
Quantity 1g
Chemical Name or Material Pyrazine-2-thiocarboxamide
MDL Number MFCD00173663
Health Hazard 1 Warning
Synonym Pyrazine-2-carbothioamide
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 4604-72-2
Odor Odorless
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Melting Point 198° to 200°C
Molecular Formula C5H5N3S

Solubility
Soluble in organic solvents, ethanol, DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.