missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrazine-2-carboxamidoxime, 97%

Manufacturer:  Alfa Aesar™ H5483406

 View more versions of this product

Catalog No. AAH5483406



Specifications

Specifications

Pyrazine-2-carboxamidoxime
51285-05-3
MFCD02258397
IXCAPURAZXLMLQ-UHFFFAOYSA-N
nitroso(1H-pyrazin-2-ylidene)methanamine
6762607
97%
5g
C5H6N4O
n'-hydroxy-2-pyrazinecarboximidamide, n'-hydroxypyrazin-2-carboximidamide, n'-hydroxypyrazine-2-carboximidamide, n-hydroxy-2-pyrazinecarboximidamide, n-hydroxy-pyrazine-2-carboxamidine, n-hydroxypyrazine-2-carboximidamide, nitroso 1h-pyrazin-2-ylidene methanamine, pyrazine-2-carboxamide oxime, qzhjcrjemifnnb-uhfffaoysa-n, z-n'-hydroxypyrazine-2-carboximidamide
C1=CN=CC(=C(N)N=O)N1
138.13
138.13

Chemical Identifiers

51285-05-3
138.13
IXCAPURAZXLMLQ-UHFFFAOYSA-N
6762607
C1=CN=CC(=C(N)N=O)N1
C5H6N4O
MFCD02258397
n'-hydroxy-2-pyrazinecarboximidamide, n'-hydroxypyrazin-2-carboximidamide, n'-hydroxypyrazine-2-carboximidamide, n-hydroxy-2-pyrazinecarboximidamide, n-hydroxy-pyrazine-2-carboxamidine, n-hydroxypyrazine-2-carboximidamide, nitroso 1h-pyrazin-2-ylidene methanamine, pyrazine-2-carboxamide oxime, qzhjcrjemifnnb-uhfffaoysa-n, z-n'-hydroxypyrazine-2-carboximidamide
nitroso(1H-pyrazin-2-ylidene)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents