missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrazine-2-carbonyl chloride, 90+%

Chemical Identifiers

CAS 19847-10-0
Molecular Formula C5H3ClN2O
Formula Weight 142.55
MDL Number MFCD00792886
UN Number UN3261
Chemical Name or Material Pyrazine-2-carbonyl chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5041803 SDS Alfa Aesar™
H5041803
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Pyrazine-2-carbonyl chloride is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with bases, oxidizing agents and moisture.
Description & Specifications

Chemical Identifiers

19847-10-0
142.55
UN3261
C5H3ClN2O
MFCD00792886
Pyrazine-2-carbonyl chloride

Specifications

1g
C5H3ClN2O
MFCD00792886
Pyrazine-2-carbonyl chloride
19847-10-0
142.55
UN3261
≥90%

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.