Learn More
Pyrazine-2-carbonitrile, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1097422
Description
Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Pyrazine-2-carbonitrile | |
18°C to 20°C | |
90°C to 93°C (15 mmHg) | |
C5H3N3 | |
MFCD00049361 | |
UN3276 | |
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
pyrazine-2-carbonitrile | |
73172 | |
105.1 |
19847-12-2 | |
1.174 | |
96°C (204°F) | |
1.536 | |
100 g | |
110008 | |
Slightly soluble in water. | |
N#CC1=CN=CC=N1 | |
105.10 | |
CHEBI:3982 | |
≥97% |
Chemical Identifiers
19847-12-2 | |
105.10 | |
PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
73172 | |
pyrazine-2-carbonitrile |
C5H3N3 | |
MFCD00049361 | |
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
CHEBI:3982 | |
N#CC1=CN=CC=N1 |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 243-369-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only