Please login to your online account to display your discounted pricing

Pyrazinamide 99%, ACROS Organics™

$79.02 - $79.02

Chemical Identifiers

Chemical Name or Material Pyrazinamide
Name Note 99%
CAS 98-96-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H5N3O
Formula Weight 123.11
MDL Number MFCD00006132
Synonym Pyrazinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC157640250

  SDS
fisher scientific acros
157640250
Plastic bottle 25g Each for $79.02

AC157640100

  SDS
thermo fisher scientific
AC157640100
Plastic bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC157641000

  SDS
fisher scientific acros
157641000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.11
Physical Form Crystalline Powder or Needles
Assay ≥98.5%
Quantity 25g
Color White
Merck Index 15, 8067
TSCA TSCA
RTECS Number UQ2275000
EINECS Number 202-717-6
Melting Point 188° to 191°C
Packaging Plastic bottle
Synonym Pyrazinecarboxamide
Infrared Spectrum Authentic
Name Note 99%
Chemical Name or Material Pyrazinamide
Assay Percent Range 99%
Molecular Formula C5H5N3O
MDL Number MFCD00006132
CAS 98-96-4