missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrazinamide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157640250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyrazinamide | |
98-96-4 | |
White | |
98.5% min. (HPLC) | |
C5H5N3O | |
25 g | |
pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat | |
IPEHBUMCGVEMRF-UHFFFAOYSA-N | |
pyrazine-2-carboxamide | |
1046 | |
123.11 | |
Crystalline Powder or Needles |
99% | |
188°C to 191°C | |
Authentic | |
Plastic bottle | |
MFCD00006132 | |
15, 8067 | |
Solubility in water: 15mg/mL. Other solubilities: 13.8mg/mL methanol- 5.7mg/mL abs. ethanol,3.8mg/mL isopropanol- 1.0mg/mL ether,0.01mg/mL isooctane- 7.4mg/mL chloroform | |
C1=CN=C(C=N1)C(=O)N | |
123.11 | |
CHEBI:45285 | |
99% |
Chemical Identifiers
98-96-4 | |
123.11 | |
IPEHBUMCGVEMRF-UHFFFAOYSA-N | |
1046 | |
pyrazine-2-carboxamide |
C5H5N3O | |
MFCD00006132 | |
pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat | |
CHEBI:45285 | |
C1=CN=C(C=N1)C(=O)N |
Safety and Handling
EINECSNumber : 202-717-6
RTECSNumber : UQ2275000
TSCA : TSCA
RUO – Research Use Only