Pyramex™ Venture II Readers Safety Glasses
Base curved lens provides full side protection without distracting seams or traditional type side shields.
Manufacturer: Pyramex™ SB1810R10
Description
- Nose buds secure the glass and prevent slipping while providing a pillow soft fit
- Fully adjustable temples - 4 size settings
- Lenses provide 99% protection against harmful UV rays
- Lenses are coated for superior scratch resistance
- Strategic placement of lens insert is out of user's direct line of vision
Specifications
For indoor applications | |
+1.00 | |
Clear | |
Reader Spectacles |
Black | |
Scratch-resistant | |
Magnifying |