Pyramex™ Venture II Readers Safety Glasses
Base curved lens provides full side protection without distracting seams or traditional type side shields.
Manufacturer:  Pyramex™ SB1820R25
 View more versions of this product
Catalog No. 19-130-3490
Description
Description
  • Nose buds secure the glass and prevent slipping while providing a pillow soft fit
  • Fully adjustable temples - 4 size settings
  • Lenses provide 99% protection against harmful UV rays
  • Lenses are coated for superior scratch resistance
  • Strategic placement of lens insert is out of user's direct line of vision
Specifications
Specifications
For outdoor applications
+2.50
Gray
Reader Spectacles
Black
Scratch-resistant
Magnifying