Please login to your online account to display your discounted pricing

Pyr3, Tocris™

Selective TRPC3 channel blocker

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Pyr3
Assay Percent Range >98%
Molecular Formula C16H11Cl3F3N3O3
Formula Weight 456.63
Synonym 1-[4-[(2, 3, 3-Trichloro-1-oxo-2-propen-1-yl)amino]phenyl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

375110

 
r & d systems
375110
10mg Each for $218.74

375150

 
r & d systems
375150
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 456.63
Quantity 10mg
Chemical Name or Material Pyr3
Assay Percent Range >98%
Molecular Formula C16H11Cl3F3N3O3
CAS 1160514-60-2
Synonym 1-[4-[(2, 3, 3-Trichloro-1-oxo-2-propen-1-yl)amino]phenyl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C