Please login to your online account to display your discounted pricing

PyOxim, 97%, ACROS Organics™

$61.84 - $61.84

Chemical Identifiers

Chemical Name or Material PyOxim
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C17H29N5O3·F6P2
Formula Weight 527.38
Synonym [Ethyl cyano(hydroxyimino)acetato-O2]tri-1-pyrrolidinylphosphonium hexafluorophosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441410050

  SDS
thermo fisher scientific
AC441410050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441410010

  SDS
thermo fisher scientific
AC441410010
1g Each for $61.84
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 527.38
Quantity 5g
Color White to off-white
Molecular Formula C17H29N5O3·F6P2
Water 1% max.
Health Hazard 3 GHS P Statement:
Do not get in eyes, on skin, or on clothing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material PyOxim
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 153433-21-7
Synonym [Ethyl cyano(hydroxyimino)acetato-O2]tri-1-pyrrolidinylphosphonium hexafluorophosphate
Health Hazard 2 GHS H Statement:
Risk of explosion if heated under confinement.
Safety and Handling

GHS P Statement:
Do not get in eyes, on skin, or on clothing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Risk of explosion if heated under confinement.