Please login to your online account to display your discounted pricing

PX 12, Tocris

Thioredoxin-1 inhibitor

$151.85 - $640.56

Chemical Identifiers

Chemical Name or Material PX 12
Assay Percent Range >99%
Molecular Formula C7H12N2S2
Formula Weight 188.31
Synonym 2-[(1-Methylpropyl)dithio]-1H-imidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

295410

 
r & d systems
RDS295410
10mg Each for $151.85

295450

 
r & d systems
RDS295450
50mg Each for $640.56
Description & Specifications

Specifications

Formula Weight 188.31
Quantity 10mg
Chemical Name or Material PX 12
Molecular Formula C7H12N2S2
Recommended Storage Store at +4°C
CAS 141400-58-0
Synonym 2-[(1-Methylpropyl)dithio]-1H-imidazole
Assay Percent Range >99%
Safety and Handling

Store at +4°C