Please login to your online account to display your discounted pricing

PVC-Coated Reagent Bottles


Make your lab safer.

Manufacturer: fisher science education  221014EMD

 View more versions of this product

Catalog No. S31232

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Closure Color White
Height (Metric) 122mm
Diameter (Metric) 122mm
Diameter (English) 4.8 in.
Closure Screw
Color Clear
Capacity (English) 4 oz.
Material Borosilicate Glass
Capacity (Metric) 125mL
Closure Size 33-430
Capacity 4 oz., 125mL
Closure Liner PTFE
Height (English) 4.8 in.
Closure Material Polypropylene