Please login to your online account to display your discounted pricing
PVC-Coated Reagent Bottles
Make your lab safer.
$144.45 - $161.05
Specifications
Closure Screw
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Closure Color Height (Metric) Price Quantity    
S31232
fisher science education
221014EMD
4 oz., 125mL 4.8 in. White 122mm Pack of 6 Each for $144.45
S31233
fisher science education
221017EMD
8 oz., 250mL 5.9 in. White 151mm Pack of 6 Each for $161.05
Description & Specifications
Specifications
Closure Screw
Color Clear
Tough coating eliminates glass scatter if broken, so cleanup is easy and safe.
  • Inert PVC coating resists strong acids, alkalis and most solvents
  • Borosilicate glass is strong and crack resistant, and no-drip pour lip offers safer, cleaner pouring
  • 4.2 and 8.45 oz. (125 and 250mL) bottles feature a special screw cap and label to help meet OSHA requirements
  • 0.6 gal. (2300mL) bottles have a finger grip
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.