Please login to your online account to display your discounted pricing

PVC-Coated Reagent Bottles


Make your lab safer.

Manufacturer: fisher science education  221017EMD

 View more versions of this product

Catalog No. S31233

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Closure Color White
Height (Metric) 151mm
Closure Size 33-430
Closure Type Screw
Diameter (Metric) 151mm
Color Clear
Closure Material Polypropylene
Diameter (English) 5.9 in.
Closure Liner PTFE
Capacity (English) 8 oz.
Material Borosilicate Glass
Capacity (Metric) 250mL
Height (English) 5.9 in.
Capacity 8 oz., 250mL