Please login to your online account to display your discounted pricing
Dynalon™ PVC Bottles
$71.90 - $71.90
Specifications
Closure Type Screw
Color Clear
Material PVC
View More Specs
Products
Catalog Number Mfr. No. Capacity Price Quantity    
S63373
dynalon™
301945
6.75 oz., 200mL Pack of 12 Each for $71.90
Case of 6 Pack for $431.35
Description & Specifications
Specifications
Closure Type Screw
Color Clear
Material PVC
A clear alternative to glass for your classroom
PVC bottles are as clear as glass without the potential hazard. All have a wide mouth with leakproof screw closure. Temperature range -20 to +80°C.