Please login to your online account to display your discounted pricing

Purvalanol B, Tocris

Cyclin-dependent kinase inhibitor

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material Purvalanol B
Assay Percent Range >99%
Molecular Formula C20H25ClN6O3
Formula Weight 432.91
Synonym (2R)-2-[[6-[(3-Chloro-4-carboxyphenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

158150

 
r & d systems
158150
50mg Each for $1,140.36

158110

 
r & d systems
158110
10mg Each for $270.88
Description & Specifications

Specifications

Formula Weight 432.91
Quantity 50mg
Chemical Name or Material Purvalanol B
CAS 212844-54-7
Molecular Formula C20H25ClN6O3
Assay Percent Range >99%
Synonym (2R)-2-[[6-[(3-Chloro-4-carboxyphenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C