Please login to your online account to display your discounted pricing

Purvalanol A, Tocris

Cyclin-dependent kinase inhibitor

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material Purvalanol A
Assay Percent Range >98%
Molecular Formula C19H25ClN6O
Formula Weight 388.9
Synonym (2R)-2-[[6-[(3-Chlorophenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

158050

 
r & d systems
158050
50mg Each for $1,140.36

158010

 
r & d systems
158010
10mg Each for $270.88
Description & Specifications

Specifications

Formula Weight 388.9
Quantity 50mg
CAS 212844-53-6
Chemical Name or Material Purvalanol A
Assay Percent Range >98%
Molecular Formula C19H25ClN6O
Synonym (2R)-2-[[6-[(3-Chlorophenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C