PURUS™ Frames for EnviroTack™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
Manufacturer: PURUS™ PF204701W00
Description
- Easily moved for cleaning
- Non-skid backing ensures frame stays in place
- Fits all PURUS™ EnviroTack™ mats
- Strong and Durable
- Non-Slip made from Polymer Coated polyester textile
- Naturally antimicrobial, mold and mildew resistant
Specifications
For use with a 457 x 1143 mm PURUS™ Biohybrid™ mats to allow for mobility and ease of cleaning | |
White | |
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications |
100% Polystyrene | |
508 x 1194 mm | |
3.175 mm |