PURUS™ PureTouch Nylon Glove Liners
PureTouch Glove Liners deliver a comfortable barrier that prevents skin irritation without diminishing tactile sensitivity. They reduce perspiration and wick away moisture during extended use of disposable or reusable gloves.
Manufacturer: PURUS™ GLFFL
Description
For use under latex, nitrile, PVC, or any disposable or reusable glove
- Low-lint, 100%15 denier nylon composition
- Sturdy and launderable for reuse
- Compatible underglove for all cleanroom environments
- Color Coded Cuffs for visual size identification
Order Info
Available in full or half-finger styles.
Compliance
Manufactured in an ISO 9002 certified facility. Meets Federal and ASTM standards for protein content. AQL 1.5.
Specifications
No | |
Full Finger | |
Nylon | |
20 pair per pack, 15 packs per case | |
Blue |
Launderable | |
For use under any disposable or reusable gloves in Pharmaceutical, Biotech, Semiconductor, Industrial, Academic, Production and Labs. Compatible underglove for all cleanroom environments. | |
Large | |
White |