PURUS™ PureTouch Nylon Glove Liners
PureTouch Glove Liners deliver a comfortable barrier that prevents skin irritation without diminishing tactile sensitivity. They reduce perspiration and wick away moisture during extended use of disposable or reusable gloves.
Manufacturer:  PURUS™ GLFFS
 View more versions of this product
Catalog No. 19-125-917
Description
Description
For use under latex, nitrile, PVC, or any disposable or reusable glove
  • Low-lint, 100%15 denier nylon composition
  • Sturdy and launderable for reuse
  • Compatible underglove for all cleanroom environments
  • Color Coded Cuffs for visual size identification
Order Info
Available in full or half-finger styles.
Compliance
Manufactured in an ISO 9002 certified facility. Meets Federal and ASTM standards for protein content. AQL 1.5.
Specifications
Specifications
White
Full Finger
Nylon
Small
Green
Ambidextrous
Launderable
For use under any disposable or reusable gloves in Pharmaceutical, Biotech, Semiconductor, Industrial, Academic, Production and Labs. Compatible underglove for all cleanroom environments.
20 pair per pack, 15 packs per case
Non-sterile
No
Yes