missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Purus™ Latex Cleanroom Paper, 22 lb.

Write, print and photocopy in cleanroom environments where the prevention of chemical exportation and particle contamination is critical.

Manufacturer:  Purus™ LCIW2010C

 View more versions of this product

Catalog No. 19-125-906


Alert:

Contains latex.


Description

Description

  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Polyethylene binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Dimensions (W × L) 21.6cm (8.5 in.) × 27.9cm (11 in.)
  • Weight 9kg (22 lb.)
  • Paper is web-cleaned on both sides prior to sheeting

Specifications

Specifications

Polyethylene
8.5 x 11 in. (216 x 279mm)
11 in.
11 in.
22 lb.
8.5 in.
Cleanroom Paper
White
For writing, printing, photocopying in cleanroom environments
279mm
279mm
9.97kg
216mm
Documents