PURUS™ Intro Mat System
The IntroMat system from PURUS™ arrives ready to use. Available in single or double frames, each holds a 24x30 mat with your choice of a PURUS™ EnviroTack™ 30-layer or 60-layer mat in a variety of colors. The frame is strong and durable with non-slip backing to ensure it remains in place.
Manufacturer:  PURUS™ PFM31503103
 View more versions of this product
Catalog No. 19-125-939
Description
Description
Arrives ready to use
  • Adhesive mats are earth-friendly containing GMO free, renewable plant based Cardia Biohybrid™ material that reduces CO2 emissions
  • Available in single or double frames made of 100% polystyrene
  • Non-skid backing is applied to the bottom of the frame to provide stability on a variety of floor surfaces
  • Frame mat can be easily relocated to high-traffic areas and removed during routine floor maintenance
Specifications
Specifications
Double White frame
61 x 76.2 cm (24 x 30 in.)
30 Sheets/Mat
Gray