missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Purple Caps

Use with empty FastPrep™ tubes. MP Biomedicals Purple Caps are screw scaps with an O-ring for use with 2mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix E tubes for prepare samples for homogenizat

Manufacturer:  MP Biomedicals™ 115064010

 View more versions of this product

Catalog No. MP115064010



Description

Description

Purple Caps fit empty FastPrep™ tubes and are used in FastPrep systems. Add sample to empty tube, add Lysing Matrix E material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample. Purple caps are also found in the Fast DNA™ Spin Kit for Soil and Fast RNA™ Pro Soil Kit.

Specifications

Specifications

Store at Room Temperature (15–30°C).
5000 Caps
Documents