missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Purple Broth Base Control
Thermo Scientific™ Remel™ Purple Broth Base Control is a control for Purple Broths.
$644.44 - $644.44
Specifications
Description | Purple Broth Base Control |
---|---|
Format | 15mm x 103mm Durham Tube |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R062756
|
Thermo Scientific™
RE062756 |
20 x 7mL |
Pack of 20 Each for $644.44
|
|
Description
Thermo Scientific™ Remel™ Purple Broth Base Control is a control for Purple Broths.
Specifications
Purple Broth Base Control | |
15mm x 103mm Durham Tube |