missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puromycin Dihydrochloride, TCI America™

Manufacturer:  TCI America P231125MG

 View more versions of this product

Catalog No. P231125MG



Specifications

Specifications

Puromycin Dihydrochloride
≥97.0% (HPLC)
MFCD00012691
CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)NC(=O)C(CC4=CC=C(C=C4)OC)N)O.Cl.Cl
544.434
544.43
Undesignated
25mg
58-58-2
C22H31Cl2N7O5
MKSVFGKWZLUTTO-ZKWLAQDLSA-N
(2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
131632508
Crystal-Powder at 20°C
174°C

Chemical Identifiers

58-58-2
544.434
MKSVFGKWZLUTTO-ZKWLAQDLSA-N
(2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
C22H31Cl2N7O5
MFCD00012691
131632508
CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)NC(=O)C(CC4=CC=C(C=C4)OC)N)O.Cl.Cl
Safety and Handling

Safety and Handling

RTECSNumber : AU7355000

TSCA : No

Recommended Storage : Freezer (-20 °C)

Documents