missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Puritan™ PurFlock™ Ultra Non-Sterile Flocked Swabs
Ideal for various rapid diagnostic tests
Supplier: Puritan™ 3306U
Description
PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultraâs high purity characteristics makes it an ideal swab for various rapid diagnostic tests.Certifications
CE marked (for medical use), ISO 9001:2008, ISO 13485:2003
Specifications
1000/Cs. | |
6.04 in. | |
PurFlock Ultra™ Flocked Swab | |
3.14 in. | |
0.69 in. | |
5mm | |
Medical | |
Sterile |
CE, ISO 9001:2008, ISO 13485:2003 | |
Buccal cell collection, forensic evidence collection, nasopharyngeal and Urethral Cell collection, rapid diagnostic testing, DNA collection and testing, collection of Bacteria or microorganisms | |
153.4mm | |
80mm | |
17.52mm | |
0.2 in. | |
Bag | |
Regular flocked |