missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ Products Cytologic Cell Scrapers

7 in. wood sample scrapers are designed to collect cytological specimens from the cervix for early diagnosis of cervical cancer

$39.29 - $55.74

Specifications

Material Wood
Product Type Wooden Scraper
Sterile No
Length (English) 7 in.
Length (Metric) 17.7cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Quantity Style Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Style Price Quantity  
14-372-10 Puritan™
2173
Scraping and collection of cervical cells for pap smears 500 Pack Ayers Pack of 500 Each for $39.29
Case of 10 Pack for $392.90
22-028697 Puritan Medical Products
2183
Scraping and collection of cervical cells for pap smears, for scraping the endocervical canal 500 Pack Double-ended Pack of 500 Each for $55.74
Case of 10 Pack for $557.40
Specifications

Description

These 7 in. wood sample scrapers are designed to collect cytological specimens from the cervix for early diagnosis of cervical cancer. The scrapers are more effective in detecting preclinical cervical cancer than a single biopsy or other related methods. Scrapers are also effective for diagnosing lesions of other areas such as the tongue, throat, vulva, and skin.

Description & Specifications

Specifications

Wood
No
17.7cm
Wooden Scraper
7 in.