missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ Histobrush™ Cytology Collection Device

Tapered Histobrush tip is gentle and reduces discomfort

Manufacturer:  Puritan™ 2199

 View more versions of this product

Catalog No. 14-960-4



Description

Description

  • Soft nylon bristles for cervical specimen collection and pap smear tests
  • Polystyrene handle that breaks when bent at 90° angle
  • Available in sterile or nonsterile formats

Cervical specimen collection, Pap Smear tests, Buccal cell collection, Veterinary use

Certifications

CE marked, ISO 9001:2008, ISO 13485:2003

Specifications

Specifications

CE, ISO 9001:2008, ISO 13485:2003
0.3cm
Polystyrene
0.75 in.
15.7cm
17.78cm
0.20 in.
Nylon
Non-sterile
0.12 in.
Cervical specimen collection, pap smear tests, buccal cell collection
6.21 in.
7 in.
1.9cm
100 Pack
0.5cm
Histobrush™ Cytology Collection Device
Documents