missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ ESK™ Sampling Kits with Pre-filled Neutralizing Buffer

For environmental surface sampling with Pre-filled Neutralizing Buffer

Manufacturer:  Puritan™ 25-83010PDBNB

 View more versions of this product

Catalog No. 14-960-224



Description

Description

  • With Pre-filled Neutralizing Buffer
  • Sterile
  • Edge-peel packaging
  • Quick-turn leak-resistant cap
  • Polyester tipped swab with polystyrene handle
  • Swab is attached to cap
  • Overall length: 4.06in.

Specifications

Specifications

ISO 9001:2008, ISO 13485:2003
0.1 in.
2.54mm
Environmental surface sampling with pre-filled neutralizing buffer
3.125 in.
4 in.
1.73cm
300 Case
Plastic
10mL
Sampling Kit
0.2 in.
5.1mm
Prefilled neutralizing buffer
0.68 in.
79.3mm
103cm
0.33 oz.
Sterile
Documents