missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Puritan™ ESK™ Sampling Kits with Pre-filled Letheen Broth
For environmental surface sampling with Letheen Broth
Supplier: Puritan™ 25-83004PDLB
Description
Intended to inactivate quaternary ammonium compounds to determine the number of bacteria present in cosmetics and other materials; For environmental surface sampling; Easy edge-peel packaging; Quick-turn leak-resistant cap ;High-quality Puritan polyester tipped swab; Swab is securely attached to cap; Pre-filled with 4ml Letheen Broth; Available in bulk case pack; Shelf life - 18 months; Made in USASpecifications
Sampling Kit | |
Sterile | |
Environmental surface sampling with letheen broth and detection of microorganisms | |
Prefilled letheen broth | |
103 cm | |
ISO 9001:2008, ISO 13485:2003 |
Plastic | |
4 mL | |
4 in. | |
0.13 oz. | |
50 Cs. |