missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ EnviroMax™ and EnviroMax Plus™ Environmental Sampling Swabs with Pointed Tips: Non-Sterile

Large tip for maximized collection and release

Manufacturer:  Puritan™ 88010PF

 View more versions of this product

Catalog No. 14-960-201



Description

Description

  • For sampling surfaces or large equipment parts
  • Quick-turn-open leak resistant cap
  • Swab attached to cap
  • Rigid foam paddle tip
  • Large tip for collection and elution
  • Pointed foam tip
  • Handle: polypropylene
  • Overall length: 6.02in. (152.91mm)

Specifications

Specifications

Non-sterile
0.12 in.
0.61 in.
1.55cm
25/Pk.
ISO 9001:2008, SO 13485:2003, FDA GMP's
0.31cm
Environmental surface sampling, perfect for spot checks, detection of microorganisms
Documents