Please login to your online account to display your discounted pricing

Purine, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC131670050

 View more versions of this product

Catalog No. AC131670050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Total base) ≥98.5% (HPLC)
Physical Form Crystalline Powder or Crystals
Formula Weight 120.11
Quantity 5g
Color Off-white to beige
Melting Point 214° to 217°C
Beilstein 26, 354
Merck Index 15, 8052
Chemical Name or Material Purine
Assay Percent Range 99%
MDL Number MFCD00079221
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C5H4N4
CAS 120-73-0

Specifications

Chemical Name or Material Purine
CAS 120-73-0
Assay ≥98.5% (Total base) ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C5H4N4
Formula Weight 120.11
MDL Number MFCD00079221