Learn More
Labconco™ Purifier™ Horizontal Clean Bench, 4ft.
When using a Labconco Purifier™ Horizontal Clean Bench, workers are protected from particulate contamination.
Specifications
Lighting Type | Fluorescent, UV |
---|---|
Height (English) | 48.4 in. |
Height (Metric) | 122.9 cm |
Plug Type | Domestic |
Width (English) | 51 in. |
Alert:
Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.
Catalog Number | Mfr. No. | Depth (Metric) Interior | Plug Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (Metric) Interior | Plug Type | Price | Quantity | ||||
13-118-121
|
Labconco™
3888421 |
66 cm | Domestic |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
13-118-120
|
Labconco™
3888420 |
53.3 cm | Domestic |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Benches direct HEPA-filtered air horizontally over the work area to provide ISO Class 5 (Class 100) conditions and protect work from particulate contamination. Applications include plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation.
- Draw room air through top of cabinet, filter out 99.99% of particles of 0.3μm and spread sterile air over work area
- Type 304 brushed stainless-steel work surface with rounded front edge
- Provide ISO 5 (Class 100) air: fewer than 100 particles 0.5μm or larger per cubic foot of air
- Angled profile is free of protrusions that could interfere with visibility or obstruct taller users
- Differential pressure across the HEPA filter is monitored via a Minihelic™ II pressure gauge
- Feature easy-to-clean, melanine-laminated hardboard interior work surface, epoxy-coated steel on exterior
- Optional factory-installed 254nm UV lamp aids in decontamination while bench is not in use
- Chrome-plated forged brass service fixture(s) with quarter-turn knobs are mounted on the right-side panel
Order Info
Overall dimensions: 34.75L x 48.5 in. H (88 x 123cm). Work surface: 19 in. D (51cm). 115V 60Hz. Shipped assembled.
Certifications
UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Compliance
ETL listed
Specifications
Fluorescent, UV | |
122.9 cm | |
51 in. |
48.4 in. | |
Domestic | |
129.5 cm |