missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Horizontal Clean Bench, 3 ft. Width

Protect worker from particulate contamination with the Labconco Purifier™ Horizontal Clean Bench.

Specifications

Height (English) 48.4 in.
Height (Metric) 122.9 cm
Plug Type North America Plug
Width (English) 38 in.
Width (Metric) 96.5 cm
View More Specs

Alert:

Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.


Products 5
Catalog Number Mfr. No. Lighting Type Hertz Price Quantity  
Catalog Number Mfr. No. Lighting Type Hertz Price Quantity  
13-118-102
Accessories View Documents Promotion Details
Labconco™
3889702
Fluorescent 50/60 Hz N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-118-101
Accessories View Documents Promotion Details
Labconco™
3889701
Fluorescent 60 Hz N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-118-110
Accessories View Documents Promotion Details
Labconco™
3889720
Fluorescent, UV 60 Hz N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-118-100
Accessories View Documents Promotion Details
Labconco™
3889700
Fluorescent 60 Hz N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-118-103
Accessories View Documents Promotion Details
Labconco™
3889703
Fluorescent 50/60 Hz N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Horizontal laminar flow
  • Double-gasket, negative pressure HEPA filter leak protection
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Disposable prefilters (2 each on 3 ft. and 4 ft. models; 3 each on 6 ft. models; 4 each on 8 ft. models)
  • Variable speed built-in motor/blower(s) with vibration isolation pads
  • Differential pressure gauge
  • Type 304 brushed stainless steel work surface
  • Tempered safety glass side panels with utility ports with iris openings
  • UV-resistant, dry powder epoxy-coated steel exterior
  • Front-mounted light and blower switches

 

Order Info

48.4 in. (123 cm) high. 115V 60Hz. Shipped assembled.

Warranty

One year warranty on parts and service

Specifications

Specifications

48.4 in.
North America Plug
96.5 cm
122.9 cm
38 in.
SDS