Labconco™ Axiom™ Class II, Type C1 Biosafety Cabinet
Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination Technology and MyLogic Operating System.
$33730.12 - $33730.12
Specifications
Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
Product Type Safety Cabinet
Sash Opening (Metric) 25.4 cm
For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Sash Opening (English) 10 in.
View More Specs
Products 1
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
30-461-1100
View Documents Promotion Details
Labconco™
304611100
North America 115 V, 60 Hz, 12 A, Domestic UV Light, Service Fixture, Vacu-Pass Portal, Base Stand 245.1 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
  • C1 designation is pending NSF approval
Specifications
Specifications
NSF International-Listed, ETL, Modified ASHRAE 110
25.4 cm
10 in.
198.6 cm
82.8 cm
Safety Cabinet
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
78.2 in.
32.6 in.
SDS
missing translation for 'documents'
missing translation for 'documents'